CH4424851875 ChemBridge Corporation

N-benzyl-1-(3-methylphenyl)methanamine hydrochloride

  • MDL number MFCD03930957
  • Cas Number : 1048640-75-0
  • Molecular Weight : 247.7631

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/6584653

Safety Information

MSDS https://www.hit2lead.com/building-blocks/6584653
Availability details for
Quantity
  • Availability
  • Add to Cart