CH4424851876 ChemBridge Corporation

N-(2-chlorobenzyl)-1-phenylethanamine hydrochloride

  • MDL number MFCD03930942
  • Cas Number : 13541-48-5
  • Molecular Weight : 282.2082

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/6585329

Safety Information

MSDS https://www.hit2lead.com/building-blocks/6585329
Availability details for
Quantity
  • Availability
  • Add to Cart