CH4424851877 ChemBridge Corporation

N-(2-methylbenzyl)-1-phenylethanamine hydrochloride

  • MDL number MFCD03930960
  • Cas Number : 1048640-81-8
  • Molecular Weight : 261.7897

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/6585868

Safety Information

MSDS https://www.hit2lead.com/sds/6585868_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart