CH4424851879 ChemBridge Corporation

(3-methylbenzyl)(tetrahydro-2-furanylmethyl)amine hydrochloride

  • MDL number MFCD03930958
  • Cas Number : 1048640-77-2
  • Molecular Weight : 241.757

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/6587957

Safety Information

MSDS https://www.hit2lead.com/building-blocks/6587957
Availability details for
Quantity
  • Availability
  • Add to Cart