CH4424851883 ChemBridge Corporation

(cyclohexylmethyl)(tetrahydro-2-furanylmethyl)amine hydrochloride

  • MDL number MFCD03904906
  • Cas Number : 1048649-17-7
  • Molecular Weight : 233.7781

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/6591031

Safety Information

MSDS https://www.hit2lead.com/building-blocks/6591031
Availability details for
Quantity
  • Availability
  • Add to Cart