CH4424851886 ChemBridge Corporation

N-(4-fluorobenzyl)-1-phenylethanamine hydrochloride

  • MDL number MFCD03930944
  • Cas Number : 1048640-51-2
  • Molecular Weight : 265.7536

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/6594097

Safety Information

MSDS https://www.hit2lead.com/sds/6594097_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart