CH4424851887 ChemBridge Corporation

N-(1-phenylethyl)-1-propanamine hydrochloride

  • MDL number MFCD03930968
  • Cas Number : 149499-66-1
  • Molecular Weight : 199.7203

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/6594188

Safety Information

MSDS https://www.hit2lead.com/building-blocks/6594188
Availability details for
Quantity
  • Availability
  • Add to Cart