CH4424851888 ChemBridge Corporation

[2-(1-cyclohexen-1-yl)ethyl](cyclopropylmethyl)amine hydrochloride

  • MDL number MFCD03930937
  • Cas Number : 1048640-45-4
  • Molecular Weight : 215.7628

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/6596931

Safety Information

MSDS https://www.hit2lead.com/building-blocks/6596931
Availability details for
Quantity
  • Availability
  • Add to Cart