CH4424851893 ChemBridge Corporation

N-(cyclopropylmethyl)cyclohexanamine hydrochloride

  • MDL number MFCD03930966
  • Cas Number : 99175-39-0
  • Molecular Weight : 189.7255

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/6599784

Safety Information

MSDS https://www.hit2lead.com/building-blocks/6599784
Availability details for
Quantity
  • Availability
  • Add to Cart