CH4424851897 ChemBridge Corporation

N-(cyclohexylmethyl)cyclohexanamine hydrochloride

  • MDL number MFCD03930934
  • Cas Number : 861372-11-4
  • Molecular Weight : 231.8052

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/6603501

Safety Information

MSDS https://www.hit2lead.com/sds/6603501_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart