CH4424851899 ChemBridge Corporation

N-(4-chlorobenzyl)-2-(1-cyclohexen-1-yl)ethanamine hydrochloride

  • MDL number MFCD03930952
  • Cas Number : 1048640-65-8
  • Molecular Weight : 286.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin RU
Product Link https://www.hit2lead.com/building-blocks/6604958

Safety Information

MSDS https://www.hit2lead.com/sds/6604958_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart