CH4424851903 ChemBridge Corporation

N-[2-(1-cyclohexen-1-yl)ethyl]-1-propanamine hydrochloride

  • MDL number MFCD03930961
  • Cas Number : 1048640-84-1
  • Molecular Weight : 203.7521

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/6606535

Safety Information

MSDS https://www.hit2lead.com/building-blocks/6606535
Availability details for
Quantity
  • Availability
  • Add to Cart