CH4424851906 ChemBridge Corporation

[2-(1-cyclohexen-1-yl)ethyl](2-methylbenzyl)amine hydrochloride

  • MDL number MFCD03930965
  • Cas Number : 1048649-07-5
  • Molecular Weight : 265.8215

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://www.hit2lead.com/building-blocks/6609574

Safety Information

msds https://www.hit2lead.com/building-blocks/6609574
Availability details for
Quantity
  • Availability
  • Add to Cart