CH4424851908 ChemBridge Corporation

N-(2,4-dichlorobenzyl)-1-phenylethanamine hydrochloride

  • MDL number MFCD03930947
  • Cas Number : 1048640-57-8
  • Molecular Weight : 316.6532

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/6612022

Safety Information

MSDS https://www.hit2lead.com/sds/6612022_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart