CH4424851910 ChemBridge Corporation

(4-methoxybenzyl)(tetrahydro-2-furanylmethyl)amine hydrochloride

  • MDL number MFCD03930962
  • Cas Number : 1048640-87-4
  • Molecular Weight : 257.7564

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/6613930

Safety Information

MSDS https://www.hit2lead.com/building-blocks/6613930
Availability details for
Quantity
  • Availability
  • Add to Cart