CH4424851912 ChemBridge Corporation

(2-phenoxyethyl)(tetrahydro-2-furanylmethyl)amine hydrochloride

  • MDL number MFCD03930936
  • Cas Number : 1048640-43-2
  • Molecular Weight : 257.7564

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/6617749

Safety Information

MSDS https://www.hit2lead.com/building-blocks/6617749
Availability details for
Quantity
  • Availability
  • Add to Cart