CH4424851934 ChemBridge Corporation

2-hydrazino-4-(4-methoxyphenyl)-1,3-thiazole hydrochloride

  • MDL number MFCD02253806
  • Cas Number : 4871-26-5
  • Molecular Weight : 257.7398

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4100610

Safety Information

MSDS https://www.hit2lead.com/sds/4100610_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart