CH4424851948 ChemBridge Corporation

N-(2-phenoxyethyl)-1,2-benzenediamine hydrochloride

  • MDL number MFCD13194084
  • Cas Number : 1609400-99-8
  • Molecular Weight : 264.7506

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/6982556

Safety Information

MSDS https://www.hit2lead.com/sds/6982556_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart