CH4424851949 ChemBridge Corporation

[1-(2-furyl)-3-buten-1-yl](2-thienylmethyl)amine hydrochloride

  • MDL number MFCD03807264
  • Cas Number : 1179367-69-1
  • Molecular Weight : 269.7902

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. -20
Physical Form solid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/6993724

Safety Information

MSDS https://www.hit2lead.com/sds/6993724_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart