CH4424851954 ChemBridge Corporation

1-[2-(4-chloro-2-methylphenoxy)ethyl]piperazine hydrochloride

  • MDL number MFCD13186360
  • Cas Number : 1609403-27-1
  • Molecular Weight : 291.2167

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/7008963

Safety Information

MSDS https://www.hit2lead.com/sds/7008963_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart