CH4424851961 ChemBridge Corporation

N-benzyl-2-(4-methoxyphenoxy)ethanamine hydrochloride

  • MDL number MFCD13186581
  • Cas Number : 1609400-45-4
  • Molecular Weight : 293.7885

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/7017116

Safety Information

MSDS https://www.hit2lead.com/sds/7017116_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart