CH4424851966 ChemBridge Corporation

5-(1-pyrrolidinylmethyl)-2-furoic acid hydrochloride

  • MDL number MFCD13194085
  • Cas Number : 1216778-27-6
  • Molecular Weight : 231.6761

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/7049370

Safety Information

MSDS https://www.hit2lead.com/sds/7049370_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart