CH4424851967 ChemBridge Corporation

2-chloro-N-(1-ethyl-3,5-dimethyl-1H-pyrazol-4-yl)acetamide hydrochloride

  • MDL number MFCD06800424
  • Cas Number : 1147231-06-8
  • Molecular Weight : 252.1409

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/7090696

Safety Information

MSDS https://www.hit2lead.com/sds/7090696_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart