CH4424851969 ChemBridge Corporation

5-[(dimethylamino)methyl]-2-furoic acid hydrochloride

  • MDL number MFCD06800602
  • Cas Number : 1185300-64-4
  • Molecular Weight : 205.6388

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/7098544

Safety Information

MSDS https://www.hit2lead.com/building-blocks/7098544
Availability details for
Quantity
  • Availability
  • Add to Cart