CH4424851972 ChemBridge Corporation

4-amino-N-(tetrahydro-2-furanylmethyl)benzamide hydrochloride

  • MDL number MFCD09763714
  • Cas Number : 1158242-72-8
  • Molecular Weight : 256.7286

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/7116375

Safety Information

MSDS https://www.hit2lead.com/sds/7116375_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart