CH4424851983 ChemBridge Corporation

N-2-propyn-1-yl-2-propen-1-amine hydrochloride

  • MDL number MFCD03596896
  • Cas Number : 1158414-22-2
  • Molecular Weight : 131.6033

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/7355693

Safety Information

MSDS https://www.hit2lead.com/building-blocks/7355693
Availability details for
Quantity
  • Availability
  • Add to Cart