CH4424851991 ChemBridge Corporation

[2-(2-bromo-4-methylphenoxy)ethyl]methylamine hydrochloride

  • MDL number MFCD26959585
  • Cas Number : 1609406-49-6
  • Molecular Weight : 280.5892

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/7719085

Safety Information

MSDS https://www.hit2lead.com/sds/7719085_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart