CH4424851993 ChemBridge Corporation

2-[3-(1-piperidinyl)propoxy]benzaldehyde hydrochloride

  • MDL number MFCD13186600
  • Cas Number : 1609400-58-9
  • Molecular Weight : 283.7937

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/7724202

Safety Information

MSDS https://www.hit2lead.com/sds/7724202_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart