CH4424852011 ChemBridge Corporation

4-[3-(4-morpholinyl)propoxy]benzaldehyde hydrochloride

  • MDL number MFCD13186611
  • Cas Number : 1609402-83-6
  • Molecular Weight : 285.7665

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/7743622

Safety Information

MSDS https://www.hit2lead.com/sds/7743622_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart