CH4424852028 ChemBridge Corporation

{[3-(4-tert-butylphenyl)-1,2,4-oxadiazol-5-yl]methyl}amine hydrochloride

  • MDL number MFCD13186616
  • Cas Number : 1609406-88-3
  • Molecular Weight : 267.7545

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/9014855

Safety Information

MSDS https://www.hit2lead.com/sds/9014855_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart