CH4424852030 ChemBridge Corporation

[(3-methyl-2-thienyl)methyl](4-pyridinylmethyl)amine hydrochloride

  • MDL number MFCD07109071
  • Cas Number : 1049689-64-6
  • Molecular Weight : 254.7789

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/9017791

Safety Information

MSDS https://www.hit2lead.com/sds/9017791_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart