CH4424852031 ChemBridge Corporation

[(2E)-3-phenyl-2-propen-1-yl](tetrahydro-2-furanylmethyl)amine hydrochloride

  • MDL number MFCD07106345
  • Cas Number : 1052514-32-5
  • Molecular Weight : 253.7677

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/9017900

Safety Information

MSDS https://www.hit2lead.com/sds/9017900_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart