CH4424852033 ChemBridge Corporation

2-[(4-pyridinylmethyl)amino]-1-butanol hydrochloride

  • MDL number MFCD07106223
  • Cas Number : 1158735-57-9
  • Molecular Weight : 216.7078

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/9019007

Safety Information

MSDS https://www.hit2lead.com/sds/9019007_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart