CH4424852042 ChemBridge Corporation

2-methyl-2-[(4-pyridinylmethyl)amino]-1-propanol hydrochloride

  • MDL number MFCD07105564
  • Cas Number : 1158213-35-4
  • Molecular Weight : 216.7078

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/9027527

Safety Information

MSDS https://www.hit2lead.com/sds/9027527_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart