CH4424852043 ChemBridge Corporation

{1-[3-(2-methylphenyl)-1,2,4-oxadiazol-5-yl]ethyl}amine hydrochloride

  • MDL number MFCD09997643
  • Cas Number : 1185294-67-0
  • Molecular Weight : 239.7014

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/9027844

Safety Information

MSDS https://www.hit2lead.com/sds/9027844_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart