CH4424852067 ChemBridge Corporation

(2-furylmethyl)(3-thienylmethyl)amine hydrochloride

  • MDL number MFCD07170393
  • Cas Number : 1158231-76-5
  • Molecular Weight : 229.7264

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/9070494

Safety Information

MSDS https://www.hit2lead.com/sds/9070494_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart