CH4424852068 ChemBridge Corporation

N-[(3-methyl-2-thienyl)methyl]cyclopropanamine hydrochloride

  • MDL number MFCD07111408
  • Cas Number : 1050214-58-8
  • Molecular Weight : 203.7322

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/9070510

Safety Information

MSDS https://www.hit2lead.com/sds/9070510_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart