CH4424852069 ChemBridge Corporation

N-(4-fluorobenzyl)-2-propen-1-amine hydrochloride

  • MDL number MFCD07107810
  • Cas Number : 1049678-31-0
  • Molecular Weight : 201.6683

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/9070515

Safety Information

MSDS https://www.hit2lead.com/sds/9070515_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart