CH4424852072 ChemBridge Corporation

N-(2-methylbenzyl)-2-propen-1-amine hydrochloride

  • MDL number MFCD07110340
  • Cas Number : 1049774-26-6
  • Molecular Weight : 197.7044

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/9070534

Safety Information

MSDS https://www.hit2lead.com/sds/9070534_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart