CH4424852073 ChemBridge Corporation

N-(2-methylbenzyl)-1-butanamine hydrochloride

  • MDL number MFCD07110016
  • Cas Number : 1049773-07-0
  • Molecular Weight : 213.7469

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/9070535

Safety Information

MSDS https://www.hit2lead.com/sds/9070535_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart