CH4424852079 ChemBridge Corporation

N-[(5-methyl-2-thienyl)methyl]cyclopentanamine hydrochloride

  • MDL number MFCD07105755
  • Cas Number : 1048664-90-9
  • Molecular Weight : 231.7853

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/9070606

Safety Information

MSDS https://www.hit2lead.com/sds/9070606_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart