CH4424852084 ChemBridge Corporation

N-(4-isopropylbenzyl)ethanamine hydrochloride

  • MDL number MFCD07169809
  • Cas Number : 1049680-95-6
  • Molecular Weight : 213.7469

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/9070625

Safety Information

MSDS https://www.hit2lead.com/sds/9070625_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart