CH4424852085 ChemBridge Corporation

tert-butyl(3-thienylmethyl)amine hydrochloride

  • MDL number MFCD07170340
  • Cas Number : 1049713-24-7
  • Molecular Weight : 205.748

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/9070637

Safety Information

MSDS https://www.hit2lead.com/sds/9070637_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart