CH4424852089 ChemBridge Corporation

(2-isobutoxybenzyl)methylamine hydrochloride

  • MDL number MFCD26959664
  • Cas Number : 1609402-94-9
  • Molecular Weight : 229.74

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.95
Salt Data HCl
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/9070650

Safety Information

MSDS https://www.hit2lead.com/sds/9070650_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart