CH4424852100 ChemBridge Corporation

N-(4-ethylbenzyl)-2-methyl-2-propanamine hydrochloride

  • MDL number MFCD07105185
  • Cas Number : 1158300-26-5
  • Molecular Weight : 227.7735

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/9070733

Safety Information

MSDS https://www.hit2lead.com/sds/9070733_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart