CH4424852102 ChemBridge Corporation

5-ethyl-4-(1-pyrrolidinylmethyl)-2-furoic acid hydrochloride

  • MDL number MFCD11506486
  • Cas Number : 1185299-46-0
  • Molecular Weight : 259.7292

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/9070743

Safety Information

MSDS https://www.hit2lead.com/building-blocks/9070743
Availability details for
Quantity
  • Availability
  • Add to Cart