CH4424852111 ChemBridge Corporation

N-(5-chloro-2-methoxybenzyl)ethanamine hydrochloride

  • MDL number MFCD07169759
  • Cas Number : 1158592-05-2
  • Molecular Weight : 236.1382

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/9070822

Safety Information

MSDS https://www.hit2lead.com/sds/9070822_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart