CH4424852115 ChemBridge Corporation

2-methyl-2-[(2-methylbenzyl)amino]-1-propanol hydrochloride

  • MDL number MFCD07109988
  • Cas Number : 1049773-02-5
  • Molecular Weight : 229.7463

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/9070834

Safety Information

MSDS https://www.hit2lead.com/sds/9070834_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart