CH4424852119 ChemBridge Corporation

N-(2-ethoxybenzyl)-1-propanamine hydrochloride

  • MDL number MFCD07107013
  • Cas Number : 1049677-99-7
  • Molecular Weight : 229.7463

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/9070845

Safety Information

MSDS https://www.hit2lead.com/sds/9070845_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart