CH4424852128 ChemBridge Corporation

2-methyl-N-[(3-methyl-2-thienyl)methyl]-1-propanamine hydrochloride

  • MDL number MFCD07107431
  • Cas Number : 1049678-10-5
  • Molecular Weight : 219.7746

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/9070908

Safety Information

MSDS https://www.hit2lead.com/sds/9070908_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart